Titanium(IV) oxide anatase, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1317-70-0
:13463-67-7
Formula:TiO₂
MDL Number:MFCD00011269
:79.88 g/mol
:M-Clarity™ quality level = MQ200
EINECS:215-280-1
Frequently Bought Together