Order Entry
Poland
ContactUsLinkComponent
Titanium(IV) oxide anatase 98+%, powder
Titanium(IV) oxide anatase 98+%, powder
Certificates
undefined
Titanium(IV) oxide anatase 98+%, powder

Specification Test Results

  • Appearance:
    White Powder
  • Assay:
    ≥98.0 %
  • Particle size:
    ≤0.05 % (residue on 44 µm sieve)
  • pH:
    7.3 to 8.3 (suspension in water)

Specification Test Results

Related Information

Frequently Bought Together