tlenek tytanu (IV), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:13463-67-7
:1317-70-0
Formula:TiO₂
MDL Number:MFCD00011269
:79.88 g/mol
Merck Index:13,09549
Boiling Pt:2900 °C (1013 hPa)
Melting Pt:1855 °C
Density:4,2 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Temperatura otoczenia
EINECS:236-675-5
Frequently Bought Together