Order Entry
Poland
ContactUsLinkComponent
 
 
Titanium(IV) chloride ≥99.9% (trace metals basis), Reagent Grade
  SIAL208566-200G
 :  Merck
 :  
 
Titanium(IV) chloride ≥99.9% (trace metals basis), Reagent Grade
  SIAL208566-200G
 :  Merck
 :  

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    200 g
  • Seq:
    3505

 

 

 :
 :189.69 g/mol
 :13,09555
 :136 °C (1013 hPa)
 :136.2 °C
 :1.72 g/cm³ (20 °C)
 :Ambient
 :231-441-9
 :1838
 :8,II
 :GHS05
 :7550-45-0
 :Cl₄Ti
 :MFCD00011267