Order Entry
Poland
ContactUsLinkComponent
Cadmium(II) iodide, Sigma-Aldrich®
Cadmium(II) iodide, Sigma-Aldrich®
Catalog # SIAL228516-50G
Supplier:  Merck
CAS Number:  
Cadmium(II) iodide, Sigma-Aldrich®
Catalog # SIAL228516-50G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :366.22 g/mol
Merck Index:12,01657
Boiling Pt:787 °C (1013 hPa)
Melting Pt:387 °C
Density:5.67 g/cm³ (20 °C)
EINECS:232-223-6
UN:2570
ADR:6.1,III
 :M-Clarity™ quality level = MQ200
 :GHS06
 :GHS08
 :GHS09
CAS Number:7790-80-9
Formula:CdI₂
MDL Number:MFCD00010920