Copper(I) oxide, Sigma-Aldrich®
Glass bottle, 25 mL
Catalog # SIAL678945-25ML
Supplier: Merck
Jump to:
Product Details & Documents
Synonyms: Cuprous oxide, Dicopper oxide
- CAS Number:1317-39-1
- Formula:Cu₂O
- MDL Number:MFCD00010974
- Molecular Weight:143.09 g/mol
- Merck Index:13,02694
- Boiling Pt:1800 °C (1013 hPa)
- Melting Pt:1232 °C
- Density:5.7…6.2 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-270-7
- ADR:9,III
- :M-Clarity™ quality level = MQ100
- UN:3077
Specifications
Specifications
- Pack type:Glass bottle
- Pk:25 mL



