Order Entry
Poland
Orders LinkContactUsLinkComponent
Copper(I) oxide, Sigma-Aldrich®

Copper(I) oxide, Sigma-Aldrich®

Irritant
Environment
Supplier:   Merck
Copper(I) oxide, Sigma-Aldrich®
Supplier:  Merck
Certificates
Copper(I) oxide, Sigma-Aldrich®

2 Options Available Below

Product Details & Documents
Synonyms:  Cuprous oxide, Dicopper oxide
  • CAS Number:1317-39-1
  • Formula:Cu₂O
  • MDL Number:MFCD00010974
  • Molecular Weight:143.09 g/mol
  • Merck Index:13,02694
  • Boiling Pt:1800 °C (1013 hPa)
  • Melting Pt:1232 °C
  • Density:5.7…6.2 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-270-7
  • UN:3077
  • ADR:9,III
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 372 zł
    372 zł
    1 * 5 g
       372 zł

  • 25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 1 290 zł
    1 290 zł
    1 * 25 g
       1 290 zł