
1,5-Dihydroxynaphthalene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:83-56-7
:
Formula:C₁₀H₈O₂
MDL Number:MFCD00003980
:160.17 g/mol
Boiling Pt:375 °C (1013 hPa)
Melting Pt:180 °C
Density:1.41 g/cm³ (20 °C)
EINECS:201-487-4
ADR:9,III
:M-Clarity™ quality level = MQ100
UN:3077
:GHS07
Storage Temperature:Ambient
Frequently Bought Together