1-Nitronaphthalene
Catalog # PROOBCR-306
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:173.17 g/mol
Merck Index:12,06711
Boiling Pt:304 °C (1013 hPa)
Melting Pt:59…61 °C
Density:1.331 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-684-5
UN:2538
ADR:4.1,III
CAS Number:86-57-7
Formula:C₁₀H₇NO₂
MDL Number:MFCD00003913