Order Entry
Poland
ContactUsLinkComponent
1-Naphthyl isothiocyanate, Sigma-Aldrich®
1-Naphthyl isothiocyanate, Sigma-Aldrich®
Catalog # SIALN4525-10G
Supplier:  Merck
CAS Number:  
1-Naphthyl isothiocyanate, Sigma-Aldrich®
Catalog # SIALN4525-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :185.25 g/mol
Merck Index:12,06498
Boiling Pt:100 °C (0.2 mmHg)
Melting Pt:59 °C
Density:1.81 g/cm³ (20 °C)
EINECS:208-990-8
UN:2923
ADR:8,III
 :M-Clarity™ quality level = MQ100
CAS Number:551-06-4
Formula:C₁₁H₇NS
MDL Number:MFCD00003882