Naphthacene, Sigma-Aldrich®
Catalog # SIAL698415-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:228.29 g/mol
Merck Index:12,06456
Boiling Pt:437 °C (1013 hPa)
Melting Pt:356 °C
Density:1.323 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:202-138-9
CAS Number:92-24-0
Formula:C₁₈H₁₂
MDL Number:MFCD00003702