
Naphthacene 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:92-24-0
:
Formula:C₁₈H₁₂
MDL Number:MFCD00003702
:228.29 g/mol
Merck Index:12,06456
Boiling Pt:437 °C (1013 hPa)
Melting Pt:>300 °C
Density:1,323 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-138-9
UN:3077
ADR:9,III
:GHS09
Specification Test Results
- Appearance:Orange Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together