Lanthanum(III) oxide, Supelco®
Supplier: Merck
Jump to:
About this item
- CAS Number:1312-81-8
- Formula:La₂O₃
- MDL Number:MFCD00011071
- Molecular Weight:325.81 g/mol
- Boiling Pt:4200 °C (1013 hPa)
- Melting Pt:2315 °C
- Density:6.51 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ100
- Storage Temperature:Ambient
- EINECS:215-200-5
2 Options Available Below
Product Details & Documents
- CAS Number:1312-81-8
- Formula:La₂O₃
- MDL Number:MFCD00011071
- Molecular Weight:325.81 g/mol
- Boiling Pt:4200 °C (1013 hPa)
- Melting Pt:2315 °C
- Density:6.51 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ100
- Storage Temperature:Ambient
- EINECS:215-200-5



