
Erbium(III) oxide ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12061-16-4
:
Formula:Er₂O₃
MDL Number:MFCD00010991
:382.52 g/mol
Merck Index:14,3646
Melting Pt:2400 °C
Density:8.64
EINECS:235-045-7
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together