
Cerium(IV) oxide ≥99.5% (REO, rare earth oxide basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1306-38-3
:
Formula:CeO₂
MDL Number:MFCD00010933
:172.11 g/mol
Merck Index:14,1989
Boiling Pt:3468 °C (1013 hPa)
Melting Pt:2600 °C
Density:7.132
EINECS:215-150-4
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together