Ytterbium(III) fluoride, Patinal®
Product Details & Documents
Synonyms: Ytterbium trifluoride, Ytterbium fluoride
- CAS Number:13760-80-0
- Formula:F₃Yb
- MDL Number:MFCD00049615
- Molecular Weight:230.04 g/mol
- Boiling Pt:2227 °C (1013 hPa)
- Melting Pt:1163.85…1179.85 °C
- Density:8.2 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:237-354-2
- UN:3288
- ADR:6.1,III
- :Density: 8.2 g/cm³ (20 °C)



