Order Entry
Poland
Orders LinkContactUsLinkComponent
Ninhydrin

Ninhydrin

Glass bottle for solids, 100 g
Catalog # APPCA0902.0100
Irritant
Supplier:   PanReac AppliChem
Ninhydrin
Catalog # APPCA0902.0100
Supplier:  PanReac AppliChem
CAS Number:  485-47-2
Ninhydrin
Ninhydrin

Glass bottle for solids, 100 g

CAS Number:  485-47-2
Price
1 140 zł
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate
  • CAS Number:485-47-2
  • Formula:C₉H₆O₄
  • MDL Number:MFCD00003791
  • Molecular Weight:178.14 g/mol
  • Merck Index:13,06582
  • Boiling Pt:351 °C (1013 hPa)
  • Melting Pt:250…258 °C
  • Density:0.862 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:207-618-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.