Iodine monochloride ACS
:7790-99-0
:
:ClI
:MFCD00011354
:162.36 g/mol
:13,05039
:97.3 °C (1013 hPa)
:26 °C
:3.2 g/cm³
:Refrigerator
:232-236-7
:1792
:8,II
:GHS05
:GHS06
- Appearance:Dark red-brown to brown to black Liquid or solid
- Assay:1.00 to 1.20 (I/Cl ratio of the Wijs solution prepared from the reagent)
- Insoluble matter:≤0.005 %
- GC:≥99.85 %
Frequently Bought Together