

Iodine monobromide for synthesis, Sigma-Aldrich®
Catalog # 8.20738.0250
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Solubility:(20 °C) decomposes
- Melting Point:37 - 40 °C
- Molar Mass:206.81 g/mol
- Boiling Point:116 °C (1013 hPa) (decomposition)
- Density:4.41 g/cm3 (20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
About this item
:
:206.81 g/mol
Merck Index:13,05038
Boiling Pt:116 °C (1013 hPa)
Melting Pt:37…40 °C
Density:4.41 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-159-9
UN:3260
ADR:8,II
:M-Clarity™ quality level = MQ200
:GHS05
:GHS07
CAS Number:7789-33-5
Formula:IBr
MDL Number:MFCD00011353