
Dihexyl disulphide tech. 85%, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10496-15-8
:
Formula:C₁₂H₂₆S₂
MDL Number:MFCD00027300
:234.47 g/mol
Boiling Pt:85…86 °C (0.5 mmHg)
Density:0.919
Flash Pt:>110 °C (230 °F)
EINECS:234-022-9
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together