Diethyl sulphide 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:352-93-2
:
Formula:C₄H₁₀S
MDL Number:MFCD00009270
Boiling Pt:90…92 °C
Melting Pt:–104 °C
Density:0.835
Molecular Weight:90.19 g/mol
Flash Pt:–10 °C (14 °F)
Merck Index:13,03885
Storage Temperature:Ambient
EINECS:206-526-9
UN:2375
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together