Dimethyl sulphoxide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:67-68-5
:
Formula:(CH₃)₂SO
MDL Number:MFCD00002089
:78.14 g/mol
Merck Index:13,03285
Boiling Pt:189 °C (1013 hPa)
Melting Pt:18.5 °C
Density:1.101 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:87 °C
Storage Temperature:Ambient
EINECS:200-664-3
Frequently Bought Together