Thionyl chloride 99.5+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7719-09-7
:
Formula:Cl₂OS
MDL Number:MFCD00011449
:118.97 g/mol
Merck Index:12,09485
Boiling Pt:76 °C (1013 hPa)
Melting Pt:–105 °C
Density:1.64 g/cm³
Storage Temperature:Ambient
EINECS:231-748-8
UN:1836
ADR:8,I
:GHS06
:GHS05
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- GC:≥99.50 %
- HPLC:≥96.0 %
Specification Test Results
Frequently Bought Together