Order Entry
Poland
ContactUsLinkComponent
 
Ammonium sulphate ≥99% ACS, Sigma-Aldrich®
  SIALA4915-25KG
 :  Merck
 :  
Ammonium sulphate ≥99% ACS, Sigma-Aldrich®
  SIALA4915-25KG
 :  Merck
 :  

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 kg
  • Seq:
    4283

 

 

 :
 :132.14 g/mol
 :13,00559
 :336…339 °C
 :1.78 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
 :Ambient
 :231-984-1
 :01-2119455044-46
 :7783-20-2
 :(NH₄)₂SO₄
 :MFCD00003391