

Ammonium hexafluorogermanate(IV) ≥99.99% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16962-47-3
:
Formula:H₈F₆GeN₂
MDL Number:MFCD00010880
:222.68 g/mol
Melting Pt:380 °C
Density:2.564
Storage Temperature:Ambient
EINECS:241-037-4
UN:3288
ADR:6.1,III
:GHS06
:GHS07
Related Information
Frequently Bought Together