
Tetrabutylphosphonium iodide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3115-66-0
:
Formula:C₁₆H₃₆IP
MDL Number:MFCD00050246
:386.34 g/mol
Melting Pt:98…100 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together