
Tetrabutylammonium tetrafluoroborate
Catalog # FLUO007316-25G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:329.27 g/mol
Melting Pt:159…162 °C
EINECS:207-058-8
:GHS07
Storage Temperature:Ambient
CAS Number:429-42-5
Formula:C₁₆H₃₆BF₄N
MDL Number:MFCD00011634