Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Trimethyl phosphate, Sigma-Aldrich®

Glass bottle for solids, 50 g
Catalog # SIAL241024-50G
Supplier:   Merck
Trimethyl phosphate, Sigma-Aldrich®
Catalog # SIAL241024-50G
Supplier:  Merck
CAS Number:  512-56-1
Trimethyl phosphate, Sigma-Aldrich®

Glass bottle for solids, 50 g

CAS Number:  512-56-1
Price
347 zł
Log in for availability
1 * 50 g
Product Details & Documents
Synonyms:  Trimethylphosphate
  • CAS Number:512-56-1
  • Formula:C₃H₉O₄P
  • MDL Number:MFCD00008348
  • Molecular Weight:140.08 g/mol
  • Boiling Pt:197.2 °C (1013 hPa)
  • Melting Pt:–46 °C
  • Density:1.21 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ200
  • Flash Pt:>150 °C
  • Storage Temperature:Ambient
  • EINECS:208-144-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications