Tributyl phosphate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: TBP, TBPA
CAS Number:126-73-8
:
Formula:(CH₃(CH₂)₃O)₃PO
MDL Number:MFCD00009436
:266.32 g/mol
Merck Index:14,9618
Boiling Pt:155…156 °C (10 mmHg)
Melting Pt:–80 °C
Density:0.977
Flash Pt:152 °C (305 °F)
Storage Temperature:Ambient
EINECS:204-800-2
:GHS08
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together