


Tributyltin methanolate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1067-52-3
:
Formula:C₁₃H₃₀OSn
MDL Number:MFCD00009419
:321.09 g/mol
Boiling Pt:124…126 °C (4 mmHg)
Density:1.120
Flash Pt:98 °C (208 °F)
EINECS:213-933-5
UN:2788
ADR:6.1,III
:GHS06
:GHS08
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together