


Tributyltin methanolate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1067-52-3
:
Formula:C₁₃H₃₀OSn
MDL Number:MFCD00009419
:321.09 g/mol
Boiling Pt:97…97.5 °C
Density:1.11 g/cm³
Flash Pt:98 °C
EINECS:213-933-5
UN:2788
ADR:6.1,II
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4726 to 1.4746 (20°C, 589 nm)
- Elemental Analysis:≥96.0 %
Specification Test Results
Frequently Bought Together