chlorotrimetylosilan do syntezy, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:75-77-4
:
Formula:C₃H₉ClSi
MDL Number:MFCD00000502
Boiling Pt:59 °C (1013 hPa)
Melting Pt:–58 °C
Density:0,8576 g/cm³ (20 °C)
Flash Pt:–18 °C
Storage Temperature:Temperatura otoczenia
EINECS:200-900-5
UN:1298
ADR:3 (8),II
:M-Clarity™ quality level = MQ200
:108.64 g/mol
REACH:01-2119457596-25
:GHS02
:GHS06
:GHS05
Specification Test Results
- Solubility:(20 °C) (rigorous decomposition)
- Melting Point:-58 °C
- Molar Mass:108.64 g/mol
- Boiling Point:57 °C (1013 hPa)
- Vapor Pressure:250 hPa (20 °C)
- Flash Point:-18 °C
- Explosion Limit:1.5 - 46 %(V)
- Density:0.86 g/cm3 (20 °C)
- Ignition Point:420 °C
- Hazard Class:3
- Hazard Class Description:Flammable Liquid
Specification Test Results
Frequently Bought Together