Tetraethyl orthosilicate (TEOS), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:78-10-4
:
Formula:Si(OC₂H₅)₄
MDL Number:MFCD00009062
:208.33 g/mol
Merck Index:13,03882
Boiling Pt:168 °C (1013 hPa)
Melting Pt:–85.5 °C
Density:0.933 g/cm³ (20 °C)
EINECS:201-083-8
UN:1292
ADR:3,III
:M-Clarity™ quality level = MQ200
Flash Pt:38 °C
:GHS02
:GHS07
Storage Temperature:Ambient
Frequently Bought Together