Tetraethyl orthosilicate (TEOS) 98%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Tetraethoxysilane
- CAS Number:78-10-4
- Formula:Si(OC₂H₅)₄
- MDL Number:MFCD00009062
- Molecular Weight:208.33 g/mol
- Merck Index:13,03882
- Boiling Pt:166…169 °C (760 mmHg)
- Melting Pt:–77 °C
- Density:0.94 g/cm³
- Flash Pt:45 °C
- Storage Temperature:Ambient
- EINECS:201-083-8
- UN:1292
- ADR:3,III
5 Options Available Below
Product Details & Documents
Synonyms: Tetraethoxysilane
- CAS Number:78-10-4
- Formula:Si(OC₂H₅)₄
- MDL Number:MFCD00009062
- Molecular Weight:208.33 g/mol
- Merck Index:13,03882
- Boiling Pt:166…169 °C (760 mmHg)
- Melting Pt:–77 °C
- Density:0.94 g/cm³
- Flash Pt:45 °C
- Storage Temperature:Ambient
- EINECS:201-083-8
- UN:1292
- ADR:3,III
Documents
Specifications for Product Family
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3820 to 1.3840 (20°C, 589 nm)
- GC:≥97.5 %
- Color scale:≤20 APHA
Product Family Options
Product Information
- Pack typePkAvailabilityPrice



