Silicon dioxide, Sigma-Aldrich®
Catalog # SIAL634867-5EA
Supplier: Merck
CAS Number:
Specifications
- Pk:5
- Seq:0005
Specifications
About this item
:
:60.08 g/mol
Merck Index:14,08493
Boiling Pt:2230 °C (1013 hPa)
Melting Pt:1719 °C
Density:2.2 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:231-545-4
CAS Number:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232