Order Entry
Poland
Orders LinkContactUsLinkComponent
Lead(II) nitrate, Sigma-Aldrich®

Lead(II) nitrate, Sigma-Aldrich®

Health
Irritant
Environment
Supplier:   Merck
Lead(II) nitrate, Sigma-Aldrich®
Supplier:  Merck
Certificates
Lead(II) nitrate, Sigma-Aldrich®

2 Options Available Below

Product Details & Documents
Synonyms:  Lead dinitrate
  • CAS Number:10099-74-8
  • Formula:Pb(NO₃)₂
  • MDL Number:MFCD00011153
  • Molecular Weight:331.21 g/mol
  • Merck Index:13,05432
  • Melting Pt:450…470 °C
  • Density:4.53 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:233-245-9
  • UN:1469
  • ADR:5.1,II
  •  :M-Clarity™ quality level = MQ100

Documents

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 656 zł
    656 zł
    1 * 10 g
       656 zł

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 2 180 zł
    2 180 zł
    1 * 50 g
       2 180 zł

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.