Order Entry
Poland
ContactUsLinkComponent
Aluminium ammonium sulphate dodecahydrate, EMSURE® for analysis, Supelco®
Irritant
Aluminium ammonium sulphate dodecahydrate, EMSURE® for analysis, Supelco®
Catalog # 1.01031.0500
Supplier:  Merck
CAS Number:  
Irritant
Aluminium ammonium sulphate dodecahydrate, EMSURE® for analysis, Supelco®
Catalog # 1.01031.0500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Solubility:
    150 g/l (20 °C)
  • Melting Point:
    93 °C
  • Molar Mass:
    453.33 g/mol
  • Bulk Density:
    700 - 800 kg/m3
  • Boiling Point:
    200 °C (1013 hPa)
  • pH:
    2.6 (100 g/l, H2O, 25 °C)

Specification Test Results

About this item

 :
 :453.33 g/mol
Merck Index:13,00325
Boiling Pt:200 °C (1013 hPa)
Melting Pt:93 °C
Density:1.64 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
EINECS:232-055-3
 :GHS07
CAS Number:7784-26-1
 :MFCD00149958
Formula:AlNH₄(SO₄)₂·12H₂O
MDL Number:MFCD00051042