Aluminium oxide, α-phase, low surface area catalyst support trimodal
Catalog # 43833.A1
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:101.96 g/mol
Storage Temperature:Ambient
EINECS:215-691-6
CAS Number:1344-28-1
Formula:Al₂O₃
MDL Number:MFCD00003424