Aluminium oxide ≥99.997% (trace metals basis), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1344-28-1
:11092-32-3
Formula:Al₂O₃
MDL Number:MFCD00003424
:101.96 g/mol
Merck Index:13,00355
Boiling Pt:2980 °C (1013 hPa)
Melting Pt:2045 °C
Density:3.94 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:215-691-6
REACH:01-2119529248-35
Frequently Bought Together