Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Aluminium oxide
Aluminium oxide catalyst support low SiO₂
Aluminium oxide catalyst support low SiO₂
:
Thermo Fisher Scientific
Aluminium oxide catalyst support low SiO₂
:
Thermo Fisher Scientific
:
1344-28-1
:
11092-32-3
:
Al₂O₃
:
MFCD00003424
:
101.96 g/mol
:
13,00355
:
2980 °C (1013 hPa)
:
2045 °C
:
3.94 g/cm³ (20 °C)
:
Ambient
:
215-691-6
:
01-2119529248-35
msds_ALFA044849
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Tetrahydrofuran, LiChrosolv® for liquid chromatography, Supelco®
: Merck
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Methanol ≥98.5%, TECHNICAL
: VWR Chemicals
Ethanol absolute ≥99.9% (by GC), EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Tetrahydrofuran stabilised, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
: Merck
2-Propanol ≥99.5% (by GC), EMPLURA®, Supelco®
: Merck
Dichloromethane ≥99.8% stabilised, PESTINORM® for pesticide residue analysis
: VWR Chemicals
Petroleum spirit, 40…65 °C, anhydrous, TECHNICAL (max. 0.01% aromatic hydrocarbons)
: VWR Chemicals
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals