Order Entry
Poland
ContactUsLinkComponent
Aluminium oxide, calcined, insulating powder
Aluminium oxide, calcined, insulating powder
Catalog # 33195.A1
CAS Number:  
Aluminium oxide, calcined, insulating powder
Catalog # 33195.A1
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :101.96 g/mol
Merck Index:13,00355
Boiling Pt:2980 °C (1013 hPa)
Melting Pt:2045 °C
Density:3.94 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-691-6
REACH:01-2119529248-35
CAS Number:1344-28-1
 :11092-32-3
Formula:Al₂O₃
MDL Number:MFCD00003424