Strontium oxide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1314-11-0
:
Formula:SrO
MDL Number:MFCD00049558
:103.62 g/mol
Merck Index:12,09009
Boiling Pt:3000 °C (1013 hPa)
Melting Pt:2430 °C
Density:4.7 g/cm³ (20 °C)
UN:3262
ADR:8,II
:M-Clarity™ quality level = MQ100
EINECS:215-219-9
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together