Calcium fluoride
Catalog # MOLE19013605-500G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:78.07 g/mol
Merck Index:13,01669
Boiling Pt:2500 °C (1013 hPa)
Melting Pt:1418 °C
Density:3.18 g/cm³ (20 °C)
EINECS:232-188-7
:GHS07
CAS Number:7789-75-5
Formula:CaF₂
MDL Number:MFCD00010907