Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Calcium fluoride
Calcium fluoride
Calcium fluoride
:
Merck
Calcium fluoride
:
Merck
:
7789-75-5
:
:
CaF₂
:
MFCD00010907
:
78.07 g/mol
:
13,01669
:
2500 °C (1013 hPa)
:
1418 °C
:
3.18 g/cm³ (20 °C)
:
232-188-7
:
GHS07
:
Ambient
MERC1.02840.9050_TDS_EN
ProductPage_102840
msds_102840
Frequently Bought Together
Mercury(II) acetate 98%
: Thermo Fisher Scientific
Carboxymethylcellulose sodium salt 1 * 1 items
: Merck
Sodium benzoate ≥99%, purified
: VWR Chemicals
Copper(II) acetate monohydrate 98.0-102.0% ACS
: Thermo Fisher Scientific