Order Entry
Poland
ContactUsLinkComponent
Magnesium perchlorate ≥85% ACS, Fluka™
Magnesium perchlorate ≥85% ACS, Fluka™
Catalog # 222283-500G
CAS Number:  
Magnesium perchlorate ≥85% ACS, Fluka™
Catalog # 222283-500G
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Identity (IR):
    complying
  • Assay of Mg:
    Min. 10.0 %
  • Loss on drying (190°C):
    Max. 8 %
  • Titrable Acid:
    Max. 0.005 meq/g
  • Titrable Base:
    Max. 0.025 meq/g
  • Water absorbing capacity:
    Min. 25 %

Specification Test Results

About this item

Synonyms: Anhydrone
 :
 :223.21 g/mol
EINECS:233-108-3
UN:1475
 :GHS03
 :GHS07
Storage Temperature:Ambient
CAS Number:10034-81-8
Formula:Cl₂MgO₈
MDL Number:MFCD00011102
Melting Pt:251 °C (dec.)
Density:2.21 g/cm³ (18 °C)
ADR:5.1 II
Merck Index:13,05702