
Lithium molybdate(VI), Sigma-Aldrich®
Catalog # SIAL400904-250G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
- Seq:3545
Specifications
About this item
:
:173.82 g/mol
:M-Clarity™ quality level = MQ100
EINECS:236-977-7
:GHS07
CAS Number:13568-40-6
Formula:Li₂MoO₄
MDL Number:MFCD00016178