Order Entry
Poland
Orders LinkContactUsLinkComponent
Sodium periodate

Sodium periodate

Glass bottle for solids, 5 g
Catalog # BKC-15
Oxidizing
Supplier:   G-Biosciences
Sodium periodate
Catalog # BKC-15
Supplier:  G-Biosciences
CAS Number:  7790-28-5
Sodium periodate

Glass bottle for solids, 5 g

CAS Number:  7790-28-5
Price
156 zł
Log in for availability
1 * 5 g
Product Details & Documents

G-Biosciences' Sodium meta -Periodate is a mild oxidizing agent that converts carbohydrates to activated active aldehydes

G-Biosciences' Sodium meta-Periodate is also suitable for oxidizing glycoproteins.

It is used in coupling to amines with cyanborohydride reduction.

Synonyms:  Sodium metaperiodate
  • CAS Number:7790-28-5
  • Formula:NaIO₄
  • MDL Number:MFCD00003534
  • Molecular Weight:213.89 g/mol
  • Merck Index:13,08714
  • Boiling Pt:300 °C (1013 hPa)
  • Melting Pt:175 °C
  • Density:3.87 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:232-197-6
  • UN:1479
  • ADR:5.1,II

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications