Cesium fluoride 99.9% (trace metals basis), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:13400-13-0
:
Formula:CsF
MDL Number:MFCD00010960
:151.9 g/mol
Boiling Pt:1251 °C (1013 hPa)
Melting Pt:680 °C
Density:4.627 g/cm³ (20 °C)
EINECS:236-487-3
ADR:8,III
:M-Clarity™ quality level = MQ200
UN:2923
:GHS06
:GHS05
:GHS08
Specification Test Results
- Appearance (Color) :White
- Appearance (Form) :Powder or Crystals or Chunk(s) or
Powder with Chunk(s) or Crystals with Chunk(s) - X-Ray Diffraction:Conforms to Structure
- Gravimetric Analysis (% Fluorine):12.0 - 13.0 %
- Purity (based On Trace Metals Analysis):99.9%
- Trace Metal Analysis :≤1500.0 ppm
Specification Test Results
Frequently Bought Together