Order Entry
Poland
Orders LinkContactUsLinkComponent
Cesium fluoride, Sigma-Aldrich®

Cesium fluoride, Sigma-Aldrich®

Toxic
Corrosive
Supplier:   Merck
Cesium fluoride, Sigma-Aldrich®
Supplier:  Merck
Certificates
Cesium fluoride, Sigma-Aldrich®

2 Options Available Below

Product Details & Documents
  • CAS Number:13400-13-0
  • Formula:CsF
  • MDL Number:MFCD00010960
  • Molecular Weight:151.9 g/mol
  • Boiling Pt:1251 °C (1013 hPa)
  • Melting Pt:680 °C
  • Density:4.627 g/cm³ (20 °C)
  • ADR:8,III
  •  :M-Clarity™ quality level = MQ100
  •  :Density: 4,627 g/cm³ (20 °C)
  • EINECS:236-487-3
  • UN:2923
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 528 zł
    528 zł
    1 * 25 g
       528 zł

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 780 zł
    780 zł
    1 * 100 g
       780 zł