Potassium dichromate, Sigma-Aldrich®
: Merck
:7778-50-9
:
:K₂Cr₂O₇
:MFCD00011367
:294.18 g/mol
:12,07790
:500 °C (1013 hPa)
:398 °C
:2.67 g/cm³ (20 °C)
:Ambient
:3087
:5.1,II
:M-Clarity™ quality level = MQ100
:231-906-6
:01-2119454792-32
:GHS03
:GHS06
:GHS05
:GHS08
:GHS09
Frequently Bought Together